Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE11-0013-41

TE 70 PWR Semi-Dry Transfer Unit

Cytiva 11-0013-41, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

packaging

pack of 1 ea

manufacturer/tradename

Cytiva 11-0013-41

availability

not available in Japan

storage temp.

room temp

General description

ECL Semi-Dry Blotters are designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amount of buffer. Units are available with and without built-in power supplies to give maximum flexibility.

The TE 70 Semi-Dry Transfer Unit is suitable for gels up to 14 × 16 cm, such as from SE 400 and SE 600 Ruby electrophoresis units or from two miniVE gels side-by-side. The TE 77 Semi-Dry Transfer Unit accepts gels up to 21 × 26 cm or four side-by-side mini-gels.

The TE 70 PWR and TE 77 PWR come with a built-in power supply and offer an automatic stopping feature. When the buffer becomes depleted, the transfer is stopped automatically, saving the transfer before overheating. This safety feature resets automatically when the power is turned off. In addition, safety is assured as the unit is limited to 30 V.

Application

Designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amount of buffer .

Features and Benefits

  • Tested for use with Amersham ECL Western blotting reagents
  • Transfer two polyacrylamide gels in a stack.
  • Long-lasting perforated platinum-titanium and stainless steel electrodes give uniform, contamination-free transfers.
  • Use low current and voltage for transfers.
  • Units are available with or without power supply.
  • Electrotransfer proteins from polyacrylamide gels in less than 1 h in newly designed, state-of-the-art units.
  • Use minimal buffer, only enough to saturate blotting papers and membranes.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL is a trademark of Cytiva

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service