western blot: 1:1,000 using non-reduced β-galactosidase
storage temp.
2-8°C
target post-translational modification
unmodified
Specificity
The antibody may be used to detect β-galactosidase expression by the lacZ reporter gene in P-element enhancer lines in Drosophila.
Immunogen
β-galactosidase from E. coli
Physical form
Lyophilized from 0.01 M phosphate buffered saline, pH 7.2.
Preparation Note
Peritoneal fluid is fractionated resulting in primarily the immunoglobulin fraction
Reconstitution
Reconstitute with 2 mL deionized water.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Stem cells reside in a particular microenvironment known as a niche. The interaction between extrinsic cues originating from the niche and intrinsic factors in stem cells determines their identity and activity. Maintenance of stem cell identity and stem cell self-renewal
Journal of experimental & clinical cancer research : CR, 31, 9-9 (2012-02-03)
Antigen-specific CD8+ cytotoxic T lymphocytes represent potent effector cells of the adaptive immune response against viruses as well as tumours. Therefore assays capable at exploring the generation and function of cytotoxic T lymphocytes represent an important objective for both clinical
Glioblastoma (GB) is the most aggressive and lethal tumour of the central nervous system (CNS). GB cells grow rapidly and display a network of projections, ultra-long tumour microtubes (TMs), that mediate cell to cell communication. GB-TMs infiltrate throughout the brain
Reactive oxygen species (ROS) are byproducts generated during normal cellular metabolism, and redox states have been shown to influence stem cell self-renewal and lineage commitment across phyla. However, the downstream effectors of ROS signaling that control stem cell behavior remain
The Journal of comparative neurology, 505(5), 586-601 (2007-10-11)
Orexin and melanin-concentrating hormone (MCH) have been implicated in mediating a variety of different behaviors. These include sleep and wakefulness, locomotion, ingestive behaviors, and fight-or-flight response, as well as anxiety- and panic-like behaviors in rodents. Despite such diversity, all these
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.