Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G4526

Sigma-Aldrich

Monosialoganglioside GM1-pyrene labeled

from bovine brain, ~95%

Synonym(s):

GM1-Pyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211

biological source

bovine brain

assay

~95%

form

solid

shipped in

ambient

Storage temp.

−20°C

Other Notes

Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum. They contain a sialated polysaccharide chain linked to ceramide through a β-glycosidic linkage. For classification of gangliosides see Svennerholm, L., et al. (eds.), Structure and Function of Gangliosides, New York, Plenum, 1980.

Preparation Note

Vacuum dried

Analysis Note

Labeled GM1 gives blue fluorescence with activation at 366 nm

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysogangliosides: synthesis and use in preparing labeled gangliosides.
G Schwarzmann et al.
Methods in enzymology, 138, 319-341 (1987-01-01)
S Sonnino et al.
Chemistry and physics of lipids, 42(1-3), 3-26 (1986-12-15)
A report is given of recent progress in the methodology for isolation of gangliosides from natural sources, for the preparation of molecular species of gangliosides homogeneous in both the oligosaccharide and ceramide portions of the molecule, for chemical manipulation and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service