Recommended Products
form
lyophilized powder
Quality Level
color
white
solubility
1% acetic acid: 1 mg/mL
UniProt accession no.
Storage temp.
−20°C
Gene Information
human ... GHRL(51738)
Amino Acid Sequence
General description
Application
Biochem/physiol Actions
Physical form
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service