Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G3291

Sigma-Aldrich

Glutamate, Caged hydrate

powder

Synonym(s):

(S)-Methyl 1-[5-(4-amino-4-carboxybutanoyl)]-7-nitroindoline-5-acetate hydrate, Caged Glutamate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O7 · xH2O
CAS Number:
Molecular Weight:
365.34 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

color

light yellow

solubility

H2O: >10 mg/mL

storage temp.

−20°C

SMILES string

O.COC(=O)Cc1cc2CCN(C(=O)CC[C@H](N)C(O)=O)c2c(c1)[N+]([O-])=O

InChI

1S/C16H19N3O7.H2O/c1-26-14(21)8-9-6-10-4-5-18(15(10)12(7-9)19(24)25)13(20)3-2-11(17)16(22)23;/h6-7,11H,2-5,8,17H2,1H3,(H,22,23);1H2/t11-;/m0./s1

InChI key

GHHRKLFCIPHBAQ-MERQFXBCSA-N

Application

Glutamate, Caged hydrate has been used:
  • as a supplement in neurobasal medium for primary culture of mouse cortical neurons[1][2]
  • as a supplemental glutamate in laboratory-prepared food (LF)[3]
  • as a component of slicing solution used for perfusing the animal in mouse brain-slice experiments[4]

Biochem/physiol Actions

Caged glutamate is a photoactivatable compound. It is active on exposure to ultraviolet light.[5]
Glutamate derivative that is resistant to hydrolysis and biologically inert prior to photolysis.

Legal Information

Sold under license from the Medical Research Council

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of small molecules rescuing fragile X syndrome phenotypes in Drosophila
Chang S, et al.
Nature chemical biology, 4(4), 256-263 (2008)
Role of mitochondrial calcium uniporter-mediated Ca2+ and iron accumulation in traumatic brain injury
Zhang LI, et al.
Journal of Cellular and Molecular Medicine, 23(4), 2995-3009 (2019)
In vitro imaging using laser photostimulation with flavoprotein autofluorescence
Theyel BB et al.
Nature Protocols, 6(4), 502-508 (2011)
Sodium aescinate provides neuroprotection in experimental traumatic brain injury via the Nrf2-ARE pathway
Zhang Li, et al.
Brain Research Bulletin, 157, 26-36 (2020)
Remote switching of cellular activity and cell signaling using light in conjunction with quantum dots
Lugo K, et al.
Biomedical Optics Express, 3(3), 447-454 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service