Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F7803

Sigma-Aldrich

Anti-phospho-Focal Adhesion Kinase (pSer732) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-phospho-FAK (pSer732)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 125 kDa

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
immunoblotting: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PTK2(5747)
mouse ... Ptk2(14083)
rat ... Ptk2(25614)

General description

Focal Adhesion Kinase (FAK) is a 125 kDa non-receptor protein tyrosine kinase that acts as a substrate for Src and is a key element of
integrin signaling. FAK plays an important role in cell spreading, differentiation, migration, cell death, and acceleration of the G1 to S phase
transition of the cell cycle. Tyrosine 397 is the autophosphorylation site of FAK, and involved in its initial activation. This phosphorylated site
binds Src family SH2 domains and the p85 subunit of PI3-Kinase, and activates cell migration and invasion.

Specificity

Also reacts with FAK-Related Non-Kinase (FRNK).

Immunogen

The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human FAK that contains serine 732. The sequence is conserved in mouse, rat and chicken.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, containing 50% glycerol, 1.0 mg/ml BSA (IgG and protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiaki V Nishimura et al.
Development (Cambridge, England), 141(18), 3540-3550 (2014-09-04)
Neuronal migration is crucial for development of the mammalian-specific six-layered cerebral cortex. Migrating neurons are known to exhibit distinct features; they form a cytoplasmic dilation, a structure specific to migrating neurons, at the proximal region of the leading process, followed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service