F6131
Fetuin from fetal bovine serum
γ-irradiated, BioXtra, suitable for cell culture
Synonym(s):
Fetuin from fetal calf serum
About This Item
Recommended Products
sterility
γ-irradiated
product line
BioXtra
form
powder
mol wt
48.4 kDa
technique(s)
cell culture | mammalian: suitable
solubility
water: 1 mg/mL, clear to slightly hazy, colorless to faintly yellow
UniProt accession no.
storage temp.
2-8°C
Gene Information
bovine ... FETUB(504615)
Looking for similar products? Visit Product Comparison Guide
General description
The composition of bovine fetuin (weight %) is polypeptide 74%, hexose 8.3%, hexosamines 5.5%, and sialic acid 8.7%.1 This product usually assays at 5-8% total sialic acid. Nearly all of this sialic acid is bound. The amount of free sialic acid is typically less than 0.3%. A method for determining free sialic acid content has been published.
Analysis Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service