F6005
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat
IgG fraction of antiserum, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352203
Recommended Products
biological source
goat
conjugate
FITC conjugate
antibody form
IgG fraction of antiserum
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
indirect immunofluorescence: 1:40
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections
Anti-Rabbit IgG (whole molecule)-FITC antibody is specific for all rabbit IgG subclasses. Goat Anti-Rabbit IgG is conjugated to Fluorescein Isothiocyanate (FITC) in an alkaline reaction, further purified to remove unbound FITC.
Anti-Rabbit IgG (whole molecule)-FITC antibody is specific for all rabbit IgG subclasses. Goat Anti-Rabbit IgG is conjugated to Fluorescein Isothiocyanate (FITC) in an alkaline reaction, further purified to remove unbound FITC.
Immunogen
Purified rabbit IgG
Application
Anti-Rabbit IgG (whole molecule)-FITC antibody may be used for immunofluorescent labeling of mouse spleen cells at a working antibody dilution of 1:40. The antibody was used for immunohistochemistry of dorsal root ganglia neurons of chicken embryos at a working antibody dilution of 1:500 using anti-neurofilament-M as primary antibody.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Debra Akin et al.
Molecular vision, 22, 40-60 (2016-02-24)
The specialized corneal epithelium requires differentiated properties, specific for its role at the anterior surface of the eye. Thus, tight maintenance of the differentiated qualities of the corneal epithelial is essential. Pinin (PNN) is an exon junction component (EJC) that
Fast rearrangement of the neuronal growth cone?s actin cytoskeleton following VEGF stimulation
Olbrich L et al
Histochemistry and Cell Biology, doi: 10-doi: 10 (2012)
Liang Zong et al.
Journal of medical genetics, 52(8), 523-531 (2015-05-20)
Auditory neuropathy spectrum disorder (ANSD) is a form of hearing loss in which auditory signal transmission from the inner ear to the auditory nerve and brain stem is distorted, giving rise to speech perception difficulties beyond that expected for the
Kip M Connor et al.
Nature medicine, 13(7), 868-873 (2007-06-26)
Many sight-threatening diseases have two critical phases, vessel loss followed by hypoxia-driven destructive neovascularization. These diseases include retinopathy of prematurity and diabetic retinopathy, leading causes of blindness in childhood and middle age affecting over 4 million people in the United
Oliver Popp et al.
mAbs, 10(2), 290-303 (2017-11-28)
Novel biotherapeutic glycoproteins, like recombinant monoclonal antibodies (mAbs) are widely used for the treatment of numerous diseases. The N-glycans attached to the constant region of an antibody have been demonstrated to be crucial for the biological efficacy. Even minor modifications
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service