F2389
Fmoc-L-phenylalanine Sasrin® resin ester
200-400 mesh
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
extent of labeling
0.3-0.8 mmol FMOC-amino acid per g resin
particle size
200-400 mesh
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-8°C
Application
For peptide synthesis. Peptides can be cleaved from the resin with 0.5-1% CF3CO2H in CH2Cl2.
Other Notes
N-Fmoc-amino acid ester of 3-methoxy-4-oxymethyl-phenoxymethylated 1% divinylbenzene cross-linked polystyrene.
Legal Information
Sasrin is a registered trademark of Bachem Bioscience Inc.
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service