Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EPI012

Sigma-Aldrich

HDAC2 IP AND ACTIVITY ASSAY KIT

Sufficient for 25 tests

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.84

storage temp.

−20°C

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service