EP0030077857
Eppendorf® ep Dualfilter T.I.P.S.® SealMax G
volume (50—1,000 μL), PCR clean, sterile, Pyrogen free, pack of 960 ea (10 racks of 96 tips)
All Photos(1)
About This Item
UNSPSC Code:
41121600
Recommended Products
material
blue
sterility
sterile
feature
DNase free
PCR clean
RNase free
filter
Pyrogen free
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Eppendorf® 0030077857
technique(s)
PCR: suitable
maximum volume
1 mL
volume
(50—1,000 μL)
General description
The violet and white filter for the ep Dualfilter T.I.P.S SealMax filter tips combine a reliable contamination protection for your pipette from your sample liquid with virtually 100 % protection from aerosols1) and biomolecules. This Dualfilter closes immediately when it comes into contact with the overpipetted sample without inhibiting valuable material. Additionally drops and splashes are retained by the white layer, the violet layer forms the highly-effective barrier against sample liquid. Further information and certification on specific sample recovery features, PCR inhibition and aerosol protection are available on your local Eppendorf website.
ep Dualfilter T.I.P.S. and ep Dualfilter T.I.P.S. SealMax are manufactured to the highest quality standards from pure, non-recycled materials under cleanroom conditions without the use of interfering slip agents, biocides or plasticizers.
Ideally suited for methods where high sensitivity and reproducibility are essential, both ep Dualfilter T.I.P.S. and ep Dualfilter T.I.P.S. SealMax are especially recommended for applications such as PCR and microbiology where preserving sample integrity and preventing cross contamination are critical.
ep Dualfilter T.I.P.S. and ep Dualfilter T.I.P.S. SealMax are manufactured to the highest quality standards from pure, non-recycled materials under cleanroom conditions without the use of interfering slip agents, biocides or plasticizers.
Ideally suited for methods where high sensitivity and reproducibility are essential, both ep Dualfilter T.I.P.S. and ep Dualfilter T.I.P.S. SealMax are especially recommended for applications such as PCR and microbiology where preserving sample integrity and preventing cross contamination are critical.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)