Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D6897

Sigma-Aldrich

2′-Deoxycytidine 3′-monophosphate ammonium salt

≥97%

Synonym(s):

3′-dCMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N3O7P · xNH3
CAS Number:
Molecular Weight:
307.20 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

assay

≥97%

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

N.NC1=NC(=O)N(C=C1)C2CC(OP(O)(O)=O)C(CO)O2

InChI

1S/C9H14N3O7P.H3N/c10-7-1-2-12(9(14)11-7)8-3-5(6(4-13)18-8)19-20(15,16)17;/h1-2,5-6,8,13H,3-4H2,(H2,10,11,14)(H2,15,16,17);1H3

InChI key

VCTPYSBTEURORB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

2′-Deoxycytidine 3′-monophosphate (3′-dCMP) and 3′-dCMPH are used as a model molecules to elucidate the mechanism(s) of the nascent stage of DNA strand breakage and to study nucleic acid base modifications by adduct formation.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F el Ghissassi et al.
Chemical research in toxicology, 8(2), 278-283 (1995-03-01)
Lipid peroxidation (LPO) products are known to interact with DNA, yielding several types of adduct with nucleobases. In this study, we demonstrate the formation of two ethenobase adducts, 1,N6-ethenoadenine and 3,N4-ethenocytosine, by reaction of LPO products with nucleic acid bases.
S F Liu et al.
Carcinogenesis, 9(8), 1401-1404 (1988-08-01)
In vitro reaction of DNA with styrene-7,8-oxide (styrene oxide) produced five adducts, as determined by 32P-postlabeling. When styrene oxide was reacted in vitro with deoxyribonucleotides, five adducts were observed from 2'-deoxyguanosine-3'-monophosphate, two from 2'-deoxyadenosine-3'-monophosphate, none from 2'-deoxythymidine-3-monophosphate or 2'-deoxycytidine-3'-monophosphate. Chromatographic
Jiande Gu et al.
Journal of the American Chemical Society, 128(4), 1250-1252 (2006-01-26)
To elucidate the mechanism of the nascent stage of DNA strand breakage by low-energy electrons, theoretical investigations of electron attachment to nucleotides have been performed by the reliably calibrated B3LYP/DZP++ approach (Chem. Rev. 2002, 102, 231). The 2'-deoxycytidine-3'-monophosphate (3'-dCMPH) and
Jiande Gu et al.
Journal of the American Chemical Society, 128(29), 9322-9323 (2006-07-20)
A detailed understanding of DNA strand breaks induced by low energy electrons (LEE) is of crucial importance for the advancement of many areas of molecular biology and medicine. To elucidate the mechanism of DNA strand breaks by LEEs, theoretical investigations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service