Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D2322

Sigma-Aldrich

Diaphorase from Clostridium kluyveri

recombinant, expressed in E. coli, Animal-component free

Synonym(s):

Diaphorase, Lipoamide Dehydrogenase, Lipoyl Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

description

Histidine-Tagged

form

solid

specific activity

>=30 units/mg protein (biuret)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Diaphorase is used in various assays to oxidate β-NADH or β-NADPH in the presence of an electron acceptor. Diaphorase from Clostridium Kluyveri has been used for semiquinone generation during benzoquinone reduction[1].

Biochem/physiol Actions

Diaphorase catalyzes the oxidation of either β-NADH or β-NADPH in the presence of an electron acceptor such as methylene blue or 2,6-dichlorophenolindophenol. Diaphorases which are specific for either β-NADH or β-NADPH are known. The pig heart enzyme of Straub seems to have native diaphorase (β-NADH specific) as well as lipoic and lipoamide dehydrogenase activities. It is reported to be a single protein. However, Massey reports that diaphorase is probably a denatured lipoamide dehydrogenase. Pre-incubation of the pig heart preparation with Cu2+ reduces the lipoamide dehydrogenase activity and proportionately increases the β-NADH diaphorase activity. In our laboratory, we have demonstrated this copper effect to some degree on the pig heart enzyme, but no appreciable effect was observed on the Clostridium kluyveri or torula yeast preparations. The lipoamide dehydrogenase:diaphorase ratio is a measure of the denaturation.

Other Notes

The name "Diaphorase" has been loosely applied to several enzymes which catalyze the oxidation of either β-NADH or β-NADPH in the presence of an electron acceptor such as methylene blue or 2,6-dichlorophenolindophenol. Many different assay procedures and "units" are used.
Diaphorases which are specific for either β-NADH or β-NADPH are known. The pig heart enzyme of Straub seems to have native diaphorase (β-NADH specific) as well as lipoic and lipoamide dehydrogenase activities. It is reported to be a single protein. However, Massey reports that "diaphorase" is probably a denatured lipoamide dehydrogenase. Pre-incubation of the pig heart preparation with Cu2+ reduces the lipoamide dehydrogenase activity and proportionately increases the β-NADH diaphorase activity. In our laboratory, we have demonstrated this copper effect to some degree on the pig heart enzyme, but no appreciable effect was observed on the Clostridium kluyveri or torula yeast preparations. The lipoamide dehydrogenase:diaphorase ratio is a measure of the denaturation.

Unit Definition

One unit of diaphorase will oxidize 1.0 micromole of beta-NADH per minute at pH 7.5 at 25 degrees C, with the corresponding reduction of the appropriate electron acceptor.

Physical form

Supplied as a lyophilized powder containing trehalose.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudio F Gonzalez et al.
The Journal of biological chemistry, 280(24), 22590-22595 (2005-04-21)
Most bacteria contain soluble quinone-reducing flavoenzymes. However, no biological benefit for this activity has previously been demonstrated. ChrR of Pseudomonas putida is one such enzyme that has also been characterized as a chromate reductase; yet we propose that it is
Alessandra Folda et al.
Archives of biochemistry and biophysics, 517(1), 30-36 (2011-11-22)
The effects of selenite and tellurite on the mammalian enzyme lipoamide dehydrogenase were compared. Selenite acts as a substrate of lipoamide dehydrogenase in a process requiring the presence of lipoamide. In contrast, tellurite is a potent inhibitor, effective in the
Gang Chen et al.
Infection and immunity, 80(9), 3194-3205 (2012-07-04)
Anaplasma phagocytophilum is a tick-borne rickettsial pathogen that provokes an acute inflammatory response during mammalian infection. The illness caused by A. phagocytophilum, human granulocytic anaplasmosis, occurs irrespective of pathogen load and results instead from host-derived immunopathology. Thus, characterizing A. phagocytophilum
Akira Ota et al.
Journal of neural transmission (Vienna, Austria : 1996), 119(11), 1327-1342 (2012-03-07)
Aripiprazole is the only atypical antipsychotic drug known to cause the phosphorylation of AMP-activated protein kinase (AMPK) in PC12 cells. However, the molecular mechanisms underlying this phosphorylation in aripiprazole-treated PC12 cells have not yet been clarified. Here, using PC12 cells
Alexandra V Kareyeva et al.
Biochimica et biophysica acta, 1817(10), 1879-1885 (2012-04-17)
The rates of NADH-supported superoxide/hydrogen peroxide production by membrane-bound bovine heart respiratory complex I, soluble pig heart dihydrolipoamide dehydrogenase (DLDH), and by accompanying operation of these enzymes in rat heart mitochondrial matrix were measured as a function of the pool

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service