Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0821

Sigma-Aldrich

DMPK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DM1PK, MDPK, MT-PK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$403.00

$403.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$403.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$403.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

3.8-5.3 nmol/min·mg

mol wt

~105 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DMPK(1760)

Biochem/physiol Actions

DMPK (or Myotonic Dystrophy Protein Kinase) is a serine-threonine kinase that is closely related to other kinases that interact with members of the Rho family of small GTPases. DMPK protein has a leucine-rich domain at the N-terminus, a serine/threonine kinase domain in the middle, and a hydrophobic region at the C-terminus. The N-terminus of DMPK plays an important role in DMPK kinase activity, and the C-terminus of DMPK determines the intracellular localization of the protein. Myotonic dystrophy type 1 is an autosomal dominant disease caused by a trinucleotide repeat-expansion, cytosine-thymine-guanine (CTG)n, in the 3′ untranslated region of DMPK.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noboru Sasagawa et al.
Journal of biochemistry, 134(4), 537-542 (2003-11-11)
We expressed human myotonic dystrophy protein kinase (DMPK) in the fission yeast Schizosaccharomyces pombe, in which the overexpression of human DMPK affects cell growth and cell shape. The human DMPK protein has a leucine-rich domain at the N-terminus, a serine/threonine
S Salvatori et al.
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology, 26(4), 235-242 (2005-09-30)
Myotonic dystrophy type 1 (DM1) is an autosomal dominant disease caused by a trinucleotide repeat-expansion, cytosine-thymine-guanine (CTG)n, in the 3' untranslated region of a gene encoding the myotonic dystrophy protein kinase (DMPK). To correlate CTG expansion and protein expression, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service