Key Documents
DBS-FCL-2
90112815, African green monkey lung, Fibroblast
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
Product Name
DBS-FCL-2, from African green monkey lung, 90112815, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
biological source
African green monkey lung
packaging
tube of 5 μg 90112815-DNA-5UG
pkg of vial of cells 90112815-1VL
growth mode
Adherent
karyotype
2n = 60, diploid
morphology
Fibroblast
receptors
Not specified
Cell Line Origin
Monkey African Green lung
Cell Line Description
Derived from 141 day old normal male African green monkey fetus. Cells are free of adventitious agents and are non-tumourigenic. Susceptible HSV, vaccinia, rhinovirus, mumps, rubella, parainfluenza 3, Coxsackie A9 and poliovirus types 1-3.
Culture Medium
EMEM (EBSS) (M2279) + 2 mM L-Glutamine (G7513) + 1% NEAA (M7145) + 10% FBS / FCS (F2442).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:3, i.e., seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37 °C. Finite lifespan, up to 12 population doublings are expected.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Legal Information
This material is cited in a US and/or other Patent and may not be used to infringe parent claims. US Patent No. 4,040,905.
Choose from one of the most recent versions:
Certificates of Analysis (COA)