C10
12022901, human colon, Epithelial
Synonym(s):
C 10, C-10
About This Item
Recommended Products
Product Name
C10, from human colon, 12022901
biological source
human colon
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
Storage temp.
−196°C
Cell Line Origin
Cell Line Description
DNA Profile
Amelogenin: X,Y
CSF1PO: 11
D13S317: 13
D16S539: 11
D5S818: 12
D7S820: 10,13
THO1: 6,9.3
TPOX: 8,9
vWA: 14,15
Culture Medium
Subculture Routine
C10 cells grow very slowly (see growth curve); following resuscitation or subculture the cells take at least 48 hours to re-attach. Cells should be left without disturbance during this time to facilitate adhesion. Centrifugation of the cells (100 g x 5 min) at resuscitation to remove DMSO improves the establishment of a viable culture. Once attached, the cells grow in discrete islands and use of trypsin or trypsin/EDTA to subculture the cells (even without knocking the flask) yields large clumps. Further disaggregation may be achieved by repeatedly pipetting the cells.
Other Notes
Disclaimer
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service