Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MTSV1-7 CE1

10081202, human breast, Cuboidal morphology at confluence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

MTSV1-7 CE1, 10081202, from human breast

biological source

human breast

growth mode

Adherent

karyotype

Not specified

morphology

Cuboidal morphology at confluence

products

Formation of disorganised structures in collagen type I gels.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Breast cancer, morphogenesis, adhesion properties

Cell Line Description

MTSV1-7 CE1 is a clonal derivative of the human mammary luminal epithelial cell line MTSV1-7 transfected with ERBB2 expression vector, enabling the over-expression of ERBB2 proto-oncogene. Cultures of MTSV1-7 were transfected with the pSV2-erbB2 construct together with the pSV2neo plasmid. (See also 1-7HB2 Sigma catalog number 10081201).

The MTSV1-7 CE1 cell line is an ideal tool for investigation of the role of the ERBB2 receptor proto-oncogene in breast cancer morphogenesis and adhesion. MTSV1-7 CE1 cells over-express the ERBB2 receptor, inducing a reduced ability to undergo morphogenesis in vitro, and reduced expression of surface adhesion molecules E-Cadherin and alpha-2 integrin. Inhibition of morphogenesis and transcription of adhesion molecules in human mammary epithelial cells can be affected by signals generated by the ERBB2 receptor, suggesting a role for ERBB2 over-expression in tumour progression and metastasis.

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 12,13
D13S317: 12
D16S539: 8,13
D5S818: 10,13
D7S820: 8,10
THO1: 7,9
TPOX: 11
vWA: 14,17

Culture Medium

DMEM + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS) + Hydrocortisone (5 μg/ml) + Bovine Insulin (10 μg/ml).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 seeding at 3 x 104 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO<sub>2</sub>; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service