Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

tsDC

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Synonym(s):

TSDC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse bone

packaging

tube of 5 μg 01081609-DNA-5UG
pkg of vial of cells 01081609-1VL

growth mode

Semi-adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Origin

Immortalised mouse dendritic cell line

DNA Profile

Not specified

Culture Medium

IDMEM (I3390) + 2 mM L-Glutamine (G7513) + 5% FBS (F2442) + 100U/ml Penicillin + 100 ug/ml Streptomycin + 0.05 mM 2ME (M6250)

Subculture Routine

Passage cells approximately once every 2 weeks. Culture at 33 °C; 5-10% CO2. Cells are of heterogenous morphology, but the line has been cloned. Some cells adhere and others grow in suspesion. Cells are detached by short incubation in PBS plus 20mM EDTA, followed by washing in culture medium. Despite the cells slow growth, they can be highly diluted -growth does not depend on a minmum cell density.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

This material is cited in US and/or other patent and may not be used to infringe patent claims. US patent No′s. 5688692, 5866759 and international patent pending PCT/GB91/00262.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service