Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8618

Sigma-Aldrich

Chondroitinase AC from Arthrobacter aurescens

aqueous glycerol solution

Synonym(s):

Chondroitin AC-Lyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

aqueous glycerol solution

specific activity

≥100 units/mg protein

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Chondroitinase AC is an eliminase that degrades chondroitin sulfates A and C, but not chondroitin sulfate B. The enzyme cleaves, via an elimination mechanism, sulfated and non-sulfated polysaccharide chains containing (1-4) linkages between hexosamines and glucuronic acid residues. The reaction yields oligosaccharide products, mainly disaccharides, containing unsaturated uronic acids that can be detected by UV spectroscopy at 232 nm.
The enzyme shows approximately equal activity with chondroitin sulfates A and C, while the activity observed with chondroitin sulfate B is approximately 7% of this value. This activity is most likely due to the presence of chondroitin sulfates A and C (10%) in the chondroitin sulfate B.

Unit Definition

One unit will liberate 1.0 μmole of 2-acetamido-2-deoxy-3-O-(β-D-gluc-4-ene-pyranosyluronic acid)-4-O-sulfo-D-galactose from chondroitin sulfate A or 1.0 μmole of 2-acetamido-2-deoxy-3-O-(β-D-gluc-4-ene-pyranosyluronic acid)-6-O-sulfo-D-galactose from chondroitin sulfate C per min at pH 6.0 at 37 °C.

Physical form

Supplied as an aqueous solution in containing 0.02M Trizma-HCl pH 7.2 and 2% glycerol

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Pojasek et al.
Biochemical and biophysical research communications, 286(2), 343-351 (2001-08-14)
Glycosaminoglycans (GAGs) are a family of complex polysaccharides involved in a diversity of biological processes, ranging from cell signaling to blood coagulation. Chondroitin sulfate (CS) and dermatan sulfate (DS) comprise a biologically important subset of GAGs. Two of the important
Studies on the enzyme chondroitinase: product structure and ion effects.
H I NAKADA et al.
Archives of biochemistry and biophysics, 94, 244-251 (1961-08-01)
Thomas N Huckerby et al.
The FEBS journal, 272(24), 6276-6286 (2005-12-13)
Chondroitin and dermatan sulfate (CS and DS) chains were isolated from bovine tracheal cartilage and pig intestinal mucosal preparations and fragmented by enzymatic methods. The oligosaccharides studied include a disaccharide and hexasaccharides from chondroitin ABC lyase digestion as well as
J Féthière et al.
Acta crystallographica. Section D, Biological crystallography, 54(Pt 2), 279-280 (1998-10-08)
Chondroitinase AC (E.C. 4.2.2.5) overexpressed in its host, Flavobacterium heparinum, was crystallized by vapor diffusion using polyethylene glycol methyl ether as precipitant. It crystallizes in the space group P43212 or its enantiomorph with a = b = 87.1 and c

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service