Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3233

Sigma-Aldrich

c-Jun Kinase substrate from rat

kinase substrate, >90% (SDS-PAGE), buffered aqueous solution

Synonym(s):

GST-c-JUN, JNK substrate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

Product Name

c-Jun Kinase substrate from rat, >90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution

recombinant

expressed in E. coli

Quality Level

assay

>90% (SDS-PAGE)

form

buffered aqueous solution

mol wt

37 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

Highly specific substrate for JNK/SAPK.

Physical form

Solution in 25 mM Tris HCl, pH 7.4, 50 mM sodium chloride, 0.5 mM EDTA, 5 mM 2-mercaptoethanol.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Dérijard et al.
Cell, 76(6), 1025-1037 (1994-03-25)
The ultraviolet (UV) response of mammalian cells is characterized by a rapid and selective increase in gene expression mediated by AP-1 and NF-kappa B. The effect on AP-1 transcriptional activity results, in part, from enhanced phosphorylation of the c-Jun NH2-terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service