Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0374

Sigma-Aldrich

Collagenase from Clostridium histolyticum

Animal component-free, ≥300 CDU/mg solid

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

essentially salt-free, lyophilized powder

quality

Animal component-free

specific activity

≥0.25 FALGPA units/mg solid
≥300 CDU/mg solid

technique(s)

cell culture | mammalian: suitable

suitability

release of physiologically active rat pancreatic islets tested

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

This product is suitable for the disaggregation of human tumor, mouse kidney, human adult and fetal brain, lung and many other epithelia tissues. It has also been shown to be effective in liver and kidney perfusion studies, digestion of pancreas, isolation of nonparenchymal rat liver cells and hepatocyte preparation. Collagenase has also been used in the preparation of arterial tissue for the study of Advanced Glycosylation End Products. This enzyme has been tested for the release of heptatocytes at a concentration of approximately 1 mg/mL. Concentrations for digestion range from 0.1 to 5 mg/mL.

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
The collagenase product is a mixture of enzymes secreted by C. histolyticum, with different products differentiated by the relative ratios of the 10-18 components found in the secreted enzymes. The main components are two collagenases, clostripain, and a neutral protease. The synergistic action of these enzymes degrade collagen and other intracellular materialThe action of both collagenase enzymes and the neutral protease is necessary for effective release of cells from tissue. Various types of collagen are the natural substrates for collagenase.

Caution

As supplied, this product is stable for one year at -20°C. There is no loss in FALGPA or protease activity in 30 days at 37°C, 50°C and -20°C. Solutions of crude collagenase are stable if frozen quickly in aliquots (at 10 mg/mL) and kept frozen at -20°C. Further freeze-thaw cycles will damage the solution. The product retains 100% activity over 7 hours when held on ice.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Preparation Note

Solutions are typically prepared at 1-2 mg/mL in TESCA buffer (containing 50 mM TES, 0.36 mM Calcium chloride, pH 7.4 at 37°C.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chara Simitzi et al.
Frontiers in bioengineering and biotechnology, 8, 816-816 (2020-08-11)
Mesenchymal stromal cells (MSC) hold significant potential for tissue engineering applications. Modular tissue engineering involves the use of cellularized "building blocks" that can be assembled via a bottom-up approach into larger tissue-like constructs. This approach emulates more closely the complexity
S Despres et al.
Comptes rendus des seances de l'Academie des sciences. Serie D, Sciences naturelles, 290(1), 41-44 (1980-01-07)
The behaviour of cariogenic Bacteria (Streptococcus mutans) is studied with regard to collagen, which represents 90% of the dentine organic matrix. Collagenase activity of cariogenic Bacteria is measured with radioactive precursors and gel electrophoresis and compared to reference bacterial collagenase
Amit P Bhole et al.
Philosophical transactions. Series A, Mathematical, physical, and engineering sciences, 367(1902), 3339-3362 (2009-08-07)
There has been great interest in understanding the methods by which collagen-based load-bearing tissue is constructed, grown and maintained in vertebrate animals. To date, the responsibility for this process has largely been placed with mesenchymal fibroblastic cells that are thought
Farhatullah Syed et al.
PloS one, 7(2), e31430-e31430 (2012-03-03)
Dupuytren's disease (DD) is a benign, fibroproliferative disease of the palmar fascia, with excessive extracellular matrix (ECM) deposition and over-production of cytokines and growth factors, resulting in digital fixed flexion contractures limiting hand function and patient quality of life. Surgical
Martin Gelbard et al.
The Journal of urology, 187(6), 2268-2274 (2012-04-17)
Collagenase Clostridium histolyticum is an investigational nonsurgical treatment for Peyronie disease. In this phase 2b, double-blind, randomized, placebo controlled study we determined the safety and efficacy of collagenase C. histolyticum and assessed a patient reported outcome questionnaire. A total of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service