Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0286

Sigma-Aldrich

Collagen-Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

saline suspension

extent of labeling

≥1 mg collagen type III per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix spacer

1 atom

capacity

≥50 units/mL binding capacity (collagenase)

storage temp.

2-8°C

Application

Collagen-agarose is an agarose conjugate in saline suspension used in affinity chromatography, protein chromatography and specialty resins. Collagen-agarose has been used in studies assessing skin tissue engineering.

Physical form

Suspension in 1.0 M NaCl containing preservative

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spencer P Lake et al.
Annals of biomedical engineering, 39(7), 1891-1903 (2011-03-19)
The mechanical role of non-fibrillar matrix and the nature of its interaction with the collagen network in soft tissues remain poorly understood, in part because of the lack of a simple experimental model system to quantify these interactions. This study's
Spencer P Lake et al.
Journal of biomedical materials research. Part A, 99(4), 507-515 (2011-09-14)
The mechanical properties of soft tissues depend on the collagen fiber network and the surrounding non-fibrillar matrix. The mechanical role of non-fibrillar material remains poorly understood. Our recent study (Lake and Barocas, Ann Biomed Eng 2011) introduced collagen-agarose co-gels as
Anne Tristan et al.
PloS one, 4(4), e5042-e5042 (2009-04-07)
Staphylococcus aureus necrotizing pneumonia is a severe disease caused by S. aureus strains carrying the Panton Valentine leukocidin (PVL) genes (lukS-PV & lukF-PV) encoded on various bacteriophages (such as phiSLT). Clinical PVL+ strains isolated from necrotizing pneumonia display an increased
Lylla Ngo et al.
Acta histochemica, 105(1), 21-28 (2003-04-02)
This study describes a novel rapid histochemical method to survey surface properties of sperm in a model system, the sea urchin. Surface properties of live and fixed sea urchin sperm of 2 species (Strongylocentrotus purpuratus and Lytechinus pictus) in seawater
A Gaspar et al.
Journal of medicine and life, 4(2), 172-177 (2011-07-22)
The aim of this study was to obtain four collagen based porous scaffolds and to assess their in vitro biocompatibility and biodegradability in order to use them for skin tissue engineering. We have prepared four variants of collagen-based biodegradable sponges

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service