Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BM0003

Sigma-Aldrich

BMS-754807

≥98% (HPLC)

Synonym(s):

(2S)-1-[4-[(5-Cyclopropyl-1H-pyrazol-3-yl)amino]pyrrolo[2,1-f][1,2,4]triazin-2-yl]-N-(6-fluoro-3-pyridinyl)-2-methyl-2-Pyrrolidinecarboxamide, BMS 754807

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24FN9O
CAS Number:
Molecular Weight:
461.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -86 to -96°, c = 1 in methanol

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

C[C@@]1(C(NC2=CC=C(F)N=C2)=O)CCCN1C(N=C3NC4=NNC(C5CC5)=C4)=NN6C3=CC=C6

InChI

1S/C23H24FN9O/c1-23(21(34)26-15-7-8-18(24)25-13-15)9-3-10-32(23)22-28-20(17-4-2-11-33(17)31-22)27-19-12-16(29-30-19)14-5-6-14/h2,4,7-8,11-14H,3,5-6,9-10H2,1H3,(H,26,34)(H2,27,28,29,30,31)/t23-/m0/s1

InChI key

LQVXSNNAFNGRAH-QHCPKHFHSA-N

Gene Information

General description

BMS-754807 is a pyrrolotriazine, which is a potent dual inhibitor of insulin-like growth factor-1 receptor (IGF-1R) and insulin receptor (InsR) tyrosine kinase. It has been reported that BMS-754807 also acts as a reversible ATP-competitive antagonist of IGF-1R by restricting the catalytic domain of the IGF-1R.[1]

Application

BMS-754807 has been used in the embryo drug treatment.[2]

Biochem/physiol Actions

BMS-754807 is an orally available and potent dual insulin-like growth factor factor-1 receptor (IGF-1R)/ insulin receptor (InsR) tyrosine kinase inhibitor that synergistically enhances antiproliferative effects of 4-hydroxytamoxifen, letrozole, and fulvestrant in MCF-7/AC-1 cells.
BMS-754807 is an orally available and potent dual insulin-like growth factor factor-1 receptor (IGF-1R)/ insulin receptor (InsR) tyrosine kinase inhibitor.

Features and Benefits

BMS-754807 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Distant Insulin Signaling Regulates Vertebrate Pigmentation through the Sheddase Bace2.
Zhang YM, et al.
Developmental Cell, 45(5), 580-594 (2018)
Joan M Carboni et al.
Molecular cancer therapeutics, 8(12), 3341-3349 (2009-12-10)
BMS-754807 is a potent and reversible inhibitor of the insulin-like growth factor 1 receptor/insulin receptor family kinases (Ki, <2 nmol/L). It is currently in phase I development for the treatment of a variety of human cancers. BMS-754807 effectively inhibits the
Adele M Musicant et al.
Cell reports, 34(8), 108768-108768 (2021-02-25)
Mucoepidermoid carcinoma (MEC) is a life-threatening salivary gland cancer that is driven primarily by a transcriptional coactivator fusion composed of cyclic AMP-regulated transcriptional coactivator 1 (CRTC1) and mastermind-like 2 (MAML2). The mechanisms by which the chimeric CRTC1/MAML2 (C1/M2) oncoprotein rewires

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service