Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B3901

Sigma-Aldrich

Tamper-evident boxes

Holds 2 x 30 mL specimen transport tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123300

material

plastic

feature

Holds 2 x 30 mL tubes

packaging

pack of 10 ea

manufacturer/tradename

Sigma-Aldrich

size

3 1/2 in. (89 mm) × 2 3/4 in. (70 mm) × 1 1/2 in. (38 mm)

Looking for similar products? Visit Product Comparison Guide

General description

For transporting forensic evidence and medical samples with "chain of custody" assurance. Boxes are white plastic, of one-piece construction. They are sealed with plastic ties and numbered locks which must be broken to open.

Packaging

Package contains 10 boxes, 50 locks, and 100 plastic ties.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service