Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B0378

Sigma-Aldrich

Brucine sulfate salt hydrate

Synonym(s):

10,11-Dimethoxystrychnine sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C23H26N2O4)2 · H2SO4
CAS Number:
Molecular Weight:
443.50 (anhydrous basis)
Beilstein/REAXYS Number:
3875567
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

mp

~185 °C (dec.) (lit.)

SMILES string

[H]O[H].OS(O)(=O)=O.[H][C@@]12CC(=O)N3c4cc(OC)c(OC)cc4[C@]56CC[N@H]7CC(=CCO1)[C@]([H])(C[C@@H]57)[C@]2([H])[C@]36[H].[H][C@@]89CC(=O)N%10c%11cc(OC)c(OC)cc%11[C@]%12%13CC[N@H]%14CC(=CCO8)[C@]([H])(C[C@@H]%12%14)[C@]9([H])[C@]%10%13[H]

InChI

1S/2C23H26N2O4.H2O4S.H2O/c2*1-27-16-8-14-15(9-17(16)28-2)25-20(26)10-18-21-13-7-19-23(14,22(21)25)4-5-24(19)11-12(13)3-6-29-18;1-5(2,3)4;/h2*3,8-9,13,18-19,21-22H,4-7,10-11H2,1-2H3;(H2,1,2,3,4);1H2/t2*13-,18-,19-,21-,22-,23+;;/m00../s1

InChI key

DYCXTLLGAUPAQK-YZWZKZGNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brucine sulfate hydrate is a sulfate salt of brucine, a major pharmacological constituent of Strychnos nux-vomica seeds, that is known for its anti-inflammatory and analgesic activities. Brucine is also used as a chiral resolving agent for the optical resolution of tertiary acetylenic alcohols into enantiomers.[1][2]

Application

Brucine sulfate salt hydrate can be used:
  • As an electron donor in the synthesis of charge transfer complexes with electron acceptors like DDQ (2,3-dichloro-5,6-dicyano-p-benzoquinone) and chloranilic acid.[3]
  • As a chiral agent or chiral dopant in the preparation of lyotropic cholesteric liquid crystalline phases.[4]

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 3

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charge transfer complexes of brucine with chloranilic acid, 2, 3-dichloro-5, 6-dicyano-1, 4-benzoquinone and tetracyanoquinodimethane: Synthesis, spectroscopic characterization and antimicrobial activity
Airabiah H, et al.
Journal of Molecular Liquids, 286(46), 110754-110754 (2019)
Effect of alkyl chain length of alcohols on cholesteric uniaxial to cholesteric biaxial phase transitions in a potassium laurate/alcohol/potassium sulfate/water/brucine lyotropic mixture: evidence of a first-order phase transition
Reis D, et al.
The Journal of Physical Chemistry B, 117(3), 942-948 (2013)
A novel brucine gel transdermal delivery system designed for anti-inflammatory and analgesic activities
Wu P, et al.
International Journal of Molecular Sciences, 18(4), 757-757 (2017)
A new optical resolution method of tertiary acetylenic alcohol utilizing complexation with brucine.
Toda F, et al.
Tetrahedron Letters, 22(46), 4669-4672 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service