Synthetic peptide directed towards the C terminal region of human SPP1
Biochem/physiol Actions
The SPP1 gene encodes an acidic matrix protein, mainly expressed in mineralized tissues, kidney and atherosclerotic vessels. This protein also contributes to several steps in the process of prostate carcinogenesis and metastasis.
Sequence
Synthetic peptide located within the following region: DSQELSKVSREFHSHEFHSHEDMLVVDPKSKEEDKHLKFRISHELDSASS
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.