Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV00019

Sigma-Aldrich

Anti-CARD12 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

116 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CARD12(58484)

General description

CARD12 is an apoptotic protein that activates caspase-1. Rabbit Anti-CARD12 antibody binds to human, mouse and rat CARD12.

Immunogen

Synthetic peptide directed towards the C terminal region of human NLRC4

Application

Rabbit Anti-CARD12 antibody can be used for immunohistochemical (4-8μg/ml, using paraffin embedded tissues) and western blot (1μg/ml) applications.

Sequence

Synthetic peptide located within the following region: QLNLAGNRVSSDGWLAFMGVFENLKQLVFFDFSTKEFLPDPALVRKLSQV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B J Geddes et al.
Biochemical and biophysical research communications, 284(1), 77-82 (2001-05-26)
The CED4/Apaf-1 family of proteins functions as critical regulators of apoptosis and NF-kappaB signaling pathways. A novel human member of this family, called CARD12, was identified that induces apoptosis when expressed in cells. CARD12 is most similar in structure to
Chafen Lu et al.
Biochemical and biophysical research communications, 331(4), 1114-1119 (2005-05-11)
CARD12 (Ipaf/Clan) is an important regulator of caspase-1 activation. It belongs to the family of the nucleotide-binding site and leucine-rich repeat (NBS-LRR) proteins. The NBS domain of the NBS-LRR proteins contains putative ATP/GTPase-specific P-loop and Mg2+-binding site motifs. However, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service