MilliporeSigma
  • Nucleotide binding to CARD12 and its role in CARD12-mediated caspase-1 activation.
Nucleotide binding to CARD12 and its role in CARD12-mediated caspase-1 activation.
Biochemical and biophysical research communications (2005-05-11)
Chafen Lu, Anlai Wang, Lin Wang, Marion Dorsch, Timothy D Ocain, Yajun Xu
ABSTRACT
CARD12 (Ipaf/Clan) is an important regulator of caspase-1 activation. It belongs to the family of the nucleotide-binding site and leucine-rich repeat (NBS-LRR) proteins. The NBS domain of the NBS-LRR proteins contains putative ATP/GTPase-specific P-loop and Mg2+-binding site motifs. However, the nucleotide-binding properties and the function of the NBS domain are unknown. We developed a nucleotide-binding assay and investigated nucleotide binding to CARD12. We find that the NBS domain of CARD12 contains a nucleotide-binding pocket with specificity for ATP/dATP. A point mutation in the P-loop (K175R) of the NBS domain abolishes ATP/dATP binding. We further demonstrate that the nucleotide-binding site is required for CARD12-mediated caspase-1 activation. CARD12 self-association and association with procaspase-1 in transfected cells were markedly decreased by the P-loop mutation K175R. Furthermore, the P-loop mutation greatly reduced caspase-1 activation-dependent proIL-1beta processing. Thus, CARD12 function is dependent on the nucleotide-binding site. Our data provide insights into the molecular mechanisms of CARD12-mediated caspase-1 activation.