Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

AMAB90664

Sigma-Aldrich

Monoclonal Anti-CTCF antibody produced in mouse

Prestige Antibodies® Powered by Atlas Antibodies, clone CL0305, purified immunoglobulin, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$555.00

$555.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$555.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$555.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CL0305, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 1 μg/mL
immunofluorescence: 2-10 μg/test (Fixation/Permeabilization: PFA/Triton X-100)
immunohistochemistry: 1:500- 1:1000

isotype

IgG1

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTCF(10664)

General description

The CCCTC-binding factor (CTCF) gene is mapped to human chromosome 16q22.1. CTCF is a transcription factor and has 11 zinc finger motifs critical for binding to genomes.

Immunogen

Recombinant protein corresponding to CCCTC-binding factor (zinc finger protein).

Sequence
QNQTDGGEVVQDVNSSVQMVMMEQLDPTLLQMKTEVMEGTVAPEAEAAVDDTQIITLQVVNMEEQPINIGELQLVQVPVPVTVPVATTSVEELQGAYENEVSKEGLAESEPMICHTLPLPEGFQVVKVGANGEVETLEQGELPPQEDP

Epitope
Binds to an epitope located within the peptide sequence NMEEQPINIGELQLV as determined by overlapping synthetic peptides.

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CCCTC-binding factor (CTCF) protein controls organization of chromatin architecture and gene expression. It regulates gene replication, recombination and splicing. CTCF regulates the gene rearrangement events during immunoglobulin synthesis. CTCF helps in the binding of cohesion protein to chromatin during cell division. Mutations in CTCF gene impacts gene regulation leading to growth retardation and developmental disorder. Mutations in the zinc finger motifs of CTCF are implicated in tumor progression.(10)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79786

Physical form

Phospate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A widely expressed transcription factor with multiple DNA sequence specificity, CTCF, is localized at chromosome segment 16q22. 1 within one of the smallest regions of overlap for common deletions in breast and prostate cancers
Filippova GN, et al.
Genes Chromosomes Cancer, 22(1), 26-36 (1998)
CTCF: an architectural protein bridging genome topology and function
Ong CT and Corces VG
Nature Reviews. Genetics, 15(4), 234-234 (2014)
De novo mutations in the genome organizer CTCF cause intellectual disability
Gregor A, et al.
American Journal of Human Genetics, 93(1), 124-131 (2013)
Defining the multivalent functions of CTCF from chromatin state and three-dimensional chromatin interactions
Lu Y, et al.
Nucleic Acids Research, 44(13), 6200-6212 (2016)
CTCF as a multifunctional protein in genome regulation and gene expression
Kim S, et al.
Experimental & Molecular Medicine, 47(6), e166-e166 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service