Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7762

Sigma-Aldrich

L-Allylglycine

≥99% (TLC), powder

Synonym(s):

(S)-(−)-2-Amino-4-pentenoic acid, L-2-Amino-4-pentenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein/REAXYS Number:
1721512
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥99% (TLC)

form

powder

optical activity

[α]25/D −37.1°, c = 4 in H2O(lit.)

color

white

solubility

H2O: 100 mg/mL

storage temp.

room temp

SMILES string

N[C@@H](CC=C)C(O)=O

InChI

1S/C5H9NO2/c1-2-3-4(6)5(7)8/h2,4H,1,3,6H2,(H,7,8)/t4-/m0/s1

InChI key

WNNNWFKQCKFSDK-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Glutamic acid decarboxylase inhibitor.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V M Abshire et al.
Neuropharmacology, 27(11), 1171-1177 (1988-11-01)
Injection of the GABA antagonist, bicuculline methiodide into the posterior hypothalamus of rats has been shown to cause marked increases in heart rate and lesser elevations in blood pressure. Allylglycine is a potent inhibitor of the synthetic enzyme for GABA
J Thomas et al.
Physiology & behavior, 49(6), 1181-1183 (1991-06-01)
The possibility that a sex difference in gamma-aminobutyric acid (GABA) activity might be an underlying cause of the previously reported sex differences in picrotoxin-induced seizures was investigated. Male and female rats were injected (IP) with the GABA antagonist, L-allylglycine (100
M Bottlaender et al.
European journal of pharmacology, 321(1), 13-17 (1997-02-19)
The influence of decreased endogenous gamma-aminobutyric acid (GABA) concentration on benzodiazepine receptor function was studied in the brain of living baboons. Positron emission tomography and the radiotracer [11C]flumazenil combined with electroencephalography were used to determine the pharmacological properties of two
M Iijima et al.
British journal of pharmacology, 171(14), 3511-3525 (2014-03-25)
Vasopressin V1B receptor antagonists may be effective for the treatment of depression and anxiety and the objective of this study was to characterize the pharmacological profiles of two newly synthesized arginine vasopressin receptor 1B (V1B receptor) antagonists, TASP0233278 and TASP0390325.
Tracie A Paine et al.
Pharmacology, biochemistry, and behavior, 135, 97-104 (2015-06-06)
Cortical GABA regulates a number of cognitive functions including attention and working memory and is dysregulated in a number of psychiatric conditions. In schizophrenia for example, changes in GABA neurons [reduced expression of glutamic acid decarboxylase (GAD), parvalbumin (PV) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service