A5336
Asparaginase−polyethylene glycol from Escherichia coli
lyophilized powder, ~70 units/mg protein
Synonym(s):
PEG-Asparaginase
About This Item
Recommended Products
form
lyophilized powder
specific activity
~70 units/mg protein
extent of labeling
≥40 mol PEG per mol protein
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Physical form
Preparation Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service