A4721
Monoclonal Anti-ARNO (Cytohesin-2) antibody produced in mouse
~2 mg/mL, clone CYT2-21, purified immunoglobulin, buffered aqueous solution
Select a Size
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
CYT2-21, monoclonal
form
buffered aqueous solution
mol wt
antigen 40 kDa
species reactivity
rat, human
concentration
~2 mg/mL
technique(s)
immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 5-10 μg/mL using a whole extract of rat brain
isotype
IgG1
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CYTH2(9266)
rat ... Cyth2(116692)
General description
Monoclonal Anti-ARNO (Cytohesin-2) reacts specifically with ARNO, and does not cross-react with other members of the cytohesin family. The antibody associates with human and rat ARNO.
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service