Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A2602

Sigma-Aldrich

Asn-Arg-Cys-Ser-Gln-Gly-Ser-Cys-Trp-Asn, Reduced

≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H67N17O16S2
Molecular Weight:
1154.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥85% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

N[C@@H](CC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CS)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CS)C(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C44H67N17O16S2/c45-21(11-32(47)65)35(68)55-23(6-3-9-51-44(49)50)37(70)60-30(18-79)42(75)59-28(16-63)40(73)56-24(7-8-31(46)64)36(69)53-14-34(67)54-27(15-62)39(72)61-29(17-78)41(74)57-25(38(71)58-26(43(76)77)12-33(48)66)10-19-13-52-22-5-2-1-4-20(19)22/h1-2,4-5,13,21,23-30,52,62-63,78-79H,3,6-12,14-18,45H2,(H2,46,64)(H2,47,65)(H2,48,66)(H,53,69)(H,54,67)(H,55,68)(H,56,73)(H,57,74)(H,58,71)(H,59,75)(H,60,70)(H,61,72)(H,76,77)(H4,49,50,51)/t21-,23-,24-,25-,26-,27-,28-,29-,30-/m0/s1

InChI key

KEQZTNKTLNAKCM-XFPWREGGSA-N

Amino Acid Sequence

Asn-Arg-Cys-Ser-Gln-Gly-Ser-Cys-Trp-Asn

Biochem/physiol Actions

The activity of the enzymes protein disulfide-isomerase (PDI) and DsbA (a periplasmic protein thiol:disulfide oxidoreductase) can be monitored with this peptide. Oxidation of this dithiol peptide to the disulfide state is accompanied by a significant change in tryptophan fluorescence emission intensity. This permits the rapid determination of the pH-dependence of the activity of both enzymes.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Frand et al.
Trends in cell biology, 10(5), 203-210 (2000-04-08)
The folding of many secretory proteins depends upon the formation of disulphide bonds. Recent advances in genetics and cell biology have outlined a core pathway for disulphide bond formation in the endoplasmic reticulum (ER) of eukaryotic cells. In this pathway
L W Ruddock et al.
The Biochemical journal, 315 ( Pt 3), 1001-1005 (1996-05-01)
A decapeptide containing two cysteine residues at positions 3 and 8 has been designed for use in monitoring the disulphide bond-forming activity of thiol:disulphide oxidoreductases. The peptide contains a tryptophan residue adjacent to one of the cysteine residues and an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service