Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93990

Sigma-Aldrich

Atto 590 alkyne

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

product line

BioReagent

assay

≥90% (HPLC)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

λ

in ethanol (with 0.1% perchloric acid)

UV absorption

λ: 593-596 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 590 is a novel fluorescent label belonging to the class of Rhodamine dyes. The dye is designed for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Characteristic features of the label are strong absorption, high fluorescence quantum yield, high thermal and photo-stability, and very little triplet formation. The dye is highly suitable for single-molecule detection applications and high-resolution microscopy.
The alkyne modification is used in the Huisgen reaction (“Click Chemistry“).

find more information here

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ons M'Saad et al.
Nature communications, 11(1), 3850-3850 (2020-08-02)
Resolving the distribution of specific proteins at the nanoscale in the ultrastructural context of the cell is a major challenge in fluorescence microscopy. We report the discovery of a new principle for an optical contrast equivalent to electron microscopy (EM)
STED nanoscopy. A glimpse into the future.
Bianchini, P.; et al.
Cell and Tissue Research, 360(1), 143-150 (2015)
Fluoreszierende Rhodamine und fluorogene Carbopyronine fur die STED-Mikroskopie lebender Zellen.
Butkevich, Alexey N.; et al.
Angewandte Chemie (International Edition in English), 128(10), 3350-3355 (2016)
Analysis of replication factories in human cells by super-resolution light microscopy.
Cseresnyes, Z.; et al.
BMC Cell Biology, 10(1), 88-88 (2009)
Fluorescence lifetime imaging with pulsed diode laser enabled stimulated emission.
Ge, J.; Kuang, C.; Lee, SS.; Kao, FJ
Optics Express, 20(27), 28216-28216 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service