Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89883

Sigma-Aldrich

D-Altronic acid lithium salt

≥98.5% (TLC)

Synonym(s):

(2S,3R,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O7 · xLi+
CAS Number:
Molecular Weight:
196.16 (free acid basis)
UNSPSC Code:
12352201
PubChem Substance ID:

assay

≥98.5% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@@H](O)[C@H](O)C(O)=O

InChI

1S/C6H12O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h2-5,7-11H,1H2,(H,12,13)/t2-,3-,4-,5+/m1/s1

InChI key

RGHNJXZEOKUKBD-AIHAYLRMSA-N

Biochem/physiol Actions

Metabolite of pentose and glucuronate interconversions, part of the D-galacturonate degradation pathway.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polygalacturonic acid metabolism in bacteria. II. Formation and metabolism of 3-deoxy-D-glycero-2, 5-hexodiulosonic acid.
J PREISS et al.
The Journal of biological chemistry, 238, 1577-1583 (1963-05-01)
J L Dreyer
European journal of biochemistry, 166(3), 623-630 (1987-08-03)
D-Altronate hydratase and D-mannonate hydratase belong to a class of Fe2+-requiring enzymes, but the function of iron in these enzymes is largely unknown. Methods are described for the convenient preparation of both these hydratases from Escherichia coli and studies related
Uronic acid metabolism in bacteria. III. Purification and properties of D-altronic acid and D-mannonic acid dehydrases in Escherichia coli.
J D SMILEY et al.
The Journal of biological chemistry, 235, 1571-1575 (1960-06-01)
Uronic acid metabolism in bacteria. II. Purification and properties of D-altronic acid and D-mannonic acid dehydrogenases in Escherichia coli.
J HICKMAN et al.
The Journal of biological chemistry, 235, 1566-1570 (1960-06-01)
Interpretation of type 2 diabetes mellitus relevant GC-MS metabolomics fingerprints by using random forests.
Huang. J.H., et al.
Analytical Methods : Advancing Methods and Applications, 5, 4883-4889 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service