Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89385

Sigma-Aldrich

Thyroglobulin from bovine thyroid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202

biological source

bovine thyroid

form

lyophilized powder

mol wt

~670 kDa
Mr ~660000

concentration

~0.6% iodine

technique(s)

bioconjugation: suitable

solubility

H2O: 20 mg/mL, hazy, yellow

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... TG(280706)

Looking for similar products? Visit Product Comparison Guide

Application

Often used as a carrier protein for the production of antibodies. Its advantage is the large number of tyrosine residues that can be used for conjugation using the diazo reaction.

Biochem/physiol Actions

Iodine-containing protein that is stored in the thyroid gland. Converted into circulating thyroxines when the thyroid is stimulated by thyroid-stimulating hormone (TSH) from the pituitary. Because its expression is unique to the thyroid, it is a sensitive indicator of recurrence or metastasis of thyroid cancer that has been treated by thyroid ablation.

Other Notes

Iodinated precursor protein of the thyroid hormones. Structure - function[1]; Structure as deduced from base complementary DNA sequence[2]
Sales restrictions may apply

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A B Rawitch et al.
The Journal of biological chemistry, 258(4), 2079-2082 (1983-02-25)
A 19-residue, thyroxine (T4)-containing peptide, Tryp-T4, has been isolated from the tryptic digest of a low molecular weight, iodine-enriched fragment derived from 19S bovine thyroglobulin. This tryptic peptide represents the only site of significant iodination in the parent polypeptide fragment.
L Mercken et al.
Nature, 316(6029), 647-651 (1985-08-15)
In mammals, an adequate supply of thyroid hormones is essential for normal growth and neurological development. The biosynthesis of thyroid hormones involves an iodinated precursor protein, thyroglobulin, which may be considered an extreme example of a pro-hormone. Thyroglobulin is a
Kouame Adou et al.
Analytical chemistry, 81(24), 10186-10192 (2009-11-21)
A charge reduction (CR) interface for electrospray ionization that permits simultaneous analysis of nanoparticle solutions by multiple detection methods was characterized. In the direct infusion configuration, a constant flow of analyte solution undergoes electrospray ionization (ESI). The charged aerosol is
Chowdhury F Miah et al.
Surgery, 156(6), 1491-1496 (2014-12-03)
The American Thyroid Association recommends lymph node mapping (LNM) ultrasonography 6-12 months after thyroidectomy for patients with papillary thyroid cancer (PTC). The yield of LNM over thyroglobulin (TG) screening is not well defined. We sought to investigate this relationship. Post
F Latrofa et al.
Clinical and experimental immunology, 178(3), 438-446 (2014-08-20)
The subclass distribution of thyroglobulin autoantibodies (TgAb) is debated, whereas their epitope pattern is restricted. Radioidine ((131)I) treatment for Graves' disease (GD) induces a rise in TgAb levels, but it is unknown whether it modifies subclass distribution and epitope pattern

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service