Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81710

Sigma-Aldrich

L-Proline

≥99.0% (NT)

Synonym(s):

(S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein/REAXYS Number:
80810
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:

assay

≥99.0% (NT)

form

crystalline powder

optical activity

[α]20/D −84.5±1°, c = 5% in H2O

impurities

≤0.3% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤0.2% loss on drying, 110 °C

color

white

mp

228 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤20 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Proline-catalyzed asymmetric Robinson annelation; Asymmetric self-condensation of β-ionylideneacetaldehyde, retinal and related compounds

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Agami et al.
Journal of the Chemical Society. Chemical Communications, 418-418 (1984)
A.E. Asato et al.
Tetrahedron Letters, 33, 3105-3105 (1992)
David Saadoun et al.
Annals of the rheumatic diseases, 73(5), 831-837 (2013-04-23)
The standard-of-care treatment of patients with hepatitis C virus (HCV)-mixed cryoglobulinemia (MC) vasculitis includes pegylated interferon α (PegIFN)-α plus ribavirin and/or rituximab. About 30-40% of patients are non-responders or relapsers to such combination. To analyse the safety and efficacy of
Michael P Manns et al.
Gastroenterology, 147(2), 366-376 (2014-04-15)
MK-5172 is an inhibitor of the hepatitis C virus (HCV) nonstructural protein 3/4A protease; MK-5172 is taken once daily and has a higher potency and barrier to resistance than licensed protease inhibitors. We investigated the efficacy and tolerability of MK-5172
Marie Couturier et al.
The Journal of biological chemistry, 288(20), 14624-14635 (2013-04-06)
The microbial deconstruction of the plant cell wall is a key biological process that is of increasing importance with the development of a sustainable biofuel industry. The glycoside hydrolase families GH5 (PaMan5A) and GH26 (PaMan26A) endo-β-1,4-mannanases from the coprophilic ascomycete

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service