Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75005

Sigma-Aldrich

FITC–DEAE–Dextran

average mol wt 150,000

Synonym(s):

FITC–DEAE–Dextran 150, Fluorescein isothiocyanate–Diethylaminoethyl–Dextran

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

synthetic

conjugate

FITC conjugate

form

powder or crystals

mol wt

average mol wt 150,000

composition

nitrogen content, 2-5%

extent of labeling

0.001-0.020 mol/mol FITC glucose

fluorescence

λex 493 nm; λem 518 nm in 0.1 M Tris pH 8.0

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunpeng Wang et al.
Molecular medicine reports, 15(1), 146-152 (2016-12-07)
The present study aimed to identify microRNAs (miRNAs) involved in regulating retinal neovascularization and retinopathy of prematurity (ROP). A total of 80 healthy C57BL/6 neonatal mice were randomly divided into the oxygen‑induced retinopathy (OIR) group (n=40), in which 7‑day‑old mice were
Giovanni S Offeddu et al.
Biomaterials, 212, 115-125 (2019-05-22)
Recent therapeutic success of large-molecule biologics has led to intense interest in assays to measure with precision their transport across the vascular endothelium and into the target tissue. Most current in vitro endothelial models show unrealistically large permeability coefficients due

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service