Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73919

Sigma-Aldrich

Atto 655 Protein Labeling Kit

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product line

BioReagent

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 647 nm; λem 680 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

This kit contains sufficient amounts of reactive dye, buffers, and protein purification sets for performing five labeling reactions (1 mg protein each) and for the subsequent purification of the labeled protein.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Breaking the Diffraction Barrier in Fluorescence Microscopy by Optical Shelving.
Bretschneider, S.; Eggeling, Chr.; Hell, S. W.
Physical Review Letters, 98(21) (2007)
Triple-Color Super-Resolution Imaging of Live Cells. Resolving Submicroscopic Receptor Organization in the Plasma Membrane.
Wilmes, S.; Staufenbiel, M.; Lisse, D.; Richter, Ch. P.; Beutel, O.; Busch, K. B. et al.
Angewandte Chemie (International Edition in English), 124(20), 4952-4955 (2012)
Simple buffers for 3D STORM microscopy.
van de Linde, S.; Kasper, R.; Heilemann, M.; Sauer, M.
Applied Physics B: Lasers and Optics, 93(4), 725-731 (2008)
PALM and STORM. Unlocking live-cell super-resolution.
Henriques, R.; Griffiths, C.; Hesper Rego, E.; Mhlanga, Musa M.
Biopolymers, 95(5), 322-331 (2011)
Direct stochastic optical reconstruction microscopy with standard fluorescent probes.
van de Linde, S.; et al.
Nature Protocols, 6(7), 991-1009 (2011)

Articles

Protein labeling kits with Atto and Tracy dyes provide easy fluorescent labeling of purified proteins, enzymes, and antibodies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service