Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68076

Sigma-Aldrich

Dextran sulfate sodium salt from Leuconostoc spp.

low sulfate content, Mw 3000-6000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201

form

powder

quality

low sulfate content

mol wt

Mw 3000-6000

composition

Sulfur Content, 8-13%

loss

≤10% loss on drying

solubility

H2O: 0.1 g/mL, clear, colorless to slightly yellow

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran.

Application

Dextran sulfate is routinely used for the selective precipitation of lipoproteins, probe hybridization membrane-immobilized DNA, the release of DNA from DNA-histone complexes, and the inhibition of RNA binding to ribosomes.

Biochem/physiol Actions

Dextran sulfate is used for the concentration of viral particles during DNA precipitation and was shown to have a marginal inhibitory effect on Micrococcus luteus and E. coli DNA polymerases. [1]

Preparation Note

Sigma dextrans are derived from Leuconostoc mesenteroides.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dextran Sulfate as a Contaminant of DNA Extracted from Concentrated Viruses and as an Inhibitor of DNA polymerases.
HITZEMAN, R.
Journal of Virology, 255-257 (1978)
Aaron H Rose et al.
Inflammatory bowel diseases, 21(9), 2005-2015 (2015-06-16)
An important role has emerged for calpain enzymes in regulating inflammation with one isoform, calpain-2, particularly important for macrophage activation. The goal of this study was to determine the therapeutic potential of a synthetic calpain-2 inhibitor, zLLY-CH2F, for colitis and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service