Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66317

Sigma-Aldrich

Detergent Screening Kit for crystallization

Synonym(s):

Crystallization Kit for Sceening of Detergents

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

storage temp.

2-8°C

General description

Similar to the Additive Screening Kit, this kit provides a range of detergents, that may be used for optimization. Detergents often interfere with the crystallization process. They are used to influence hydrophobic sample-sample interactions which may cause non-specific aggregation. On the other hand they can also influence the structure of water. The use of detergents is typically required for crystallization of membrane proteins or other proteins, if non-specific aggregation is suspected.

The detergent kit includes 48 defined detergents, that are pre-formulated for ease of use. The concentration of each detergent solution is adjusted to the 10-fold value of critical micelle concentration. The kit includes 100 μl of each pre-formulated detergent.

Kit Components Only

Product No.
Description

  • Hexyl-β-D-glucoside 200 mM solution

  • Heptyl-β-D-glucopyranoside 150 mM solution

  • Nonyl-β-D-1-thiomaltoside solution

  • Octyl-α/β-glucoside 10 mM solution

  • Nonyl-β-D-glucoside 50 mM Solution, 50 mM

  • O-(Decylphosphoryl)choline solution

  • Methyl 6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside 200 mM solution, 200 mM

  • Triethyleneglycol monooctyl ether 1 mM solution

  • Triethyleneglycol monodecyl ether 1 mM solution

  • Tetraethyleneglycol monooctyl ether 1 mM solution

  • Tetraethyleneglycol monodecyl ether 1 mM solution

  • Tetraethyleneglycol monododecyl ether 1 mM solution

  • Pentaethyleneglycol monodecyl ether 1 mM solution

  • Pentaethyleneglycol monododecyl ether 1 mM solution

  • Hexaethyleneglycol monodecyl ether 1 mM solution

  • Hexaethyleneglycol monododecyl ether 1 mM solution

  • Heptaethyleneglycol monodecyl ether 1 mM solution

  • Heptaethyleneglycol monododecyl ether 1 mM solution

  • Octaethyleneglycol monodecyl ether 1 mM solution

  • Octaethyleneglycol monododecyl ether 1 mM solution

  • Nonaethyleneglycol monododecyl ether 1 mM solution

  • Decyl-β-D-maltoside 10 mM solution

  • Undecyl β-D-maltoside solution, 10 mM

  • Dodecyl β-D-maltoside 3 mM solution

  • Hexadecyl-β-D-maltosid 0.1 mM solution

  • Cyclohexylmethyl-β-D-maltoside 500 mM solution

  • 2-Cyclohexylethyl β-D-maltoside 200 mM solution

  • 5-Cyclohexylpentyl-β-D-maltoside 30 mM solution

  • 6-Cyclohexylhexyl-β-D-maltoside 30 mM solution

  • Heptyl-β-D-1-thioglucoside 300 mM solution

  • MEGA-8 500 mM solution

  • MEGA-9 200 mM solution

  • Decyl-β-D-1-thiomaltopyranoside solution, 10 mM

  • N,N-Dimethylhexylamine N-oxide 100 mM solution

  • O-(Tetradecylphosphoryl)choline 1.5 mM solution, 1.5 mM in H2O

  • N,N-Dimethyloctylamine N-oxide 100 mM solution

  • N,N-Dimethylnonylamine N-oxide 100 mM solution

  • N,N-Dimethyldecylamine N-oxide 100 mM solution

  • N,N-Dimethyldodecylamine N-oxide 100 mM solution

  • 3-(N,N-Dimethyloctylammonio)propanesulfonate 500 mM solution

  • 3-(N,N-Dimethyldecylammonio)propanesulfonate 300 mM solution

  • N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate 30 mM solution, 30 mM

  • 3-(N,N-Dimethylmyristylammonio)propanesulfonate 3 mM solution

  • 3-(N,N-Dimethylpalmitylammonio)propanesulfonate 0.3 mM solution

See All (44)

Kit Components Also Available Separately

Product No.
Description
SDS

  • 18842Octyl-β-D-glucopyranoside 100 mM solutionSDS

  • Triethyleneglycol monododecyl ether 1 mM solution

  • 38544O-(Octylphosphoryl)choline solution, 1 M in H2OSDS

  • 19899CHAPS 100 mM solutionSDS


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service