Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51915

Sigma-Aldrich

L-Arabinonic acid lithium salt

≥98.0% (TLC)

Synonym(s):

L-Arabonic acid lithium salt, Lithium L-arabinonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O6 · xLi+
CAS Number:
Molecular Weight:
166.13 (free acid basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98.0% (TLC)

form

solid

storage temp.

2-8°C

SMILES string

OC[C@H](O)[C@H](O)[C@@H](O)C(O)=O

InChI

1S/C5H10O6/c6-1-2(7)3(8)4(9)5(10)11/h2-4,6-9H,1H2,(H,10,11)/t2-,3-,4+/m0/s1

InChI key

QXKAIJAYHKCRRA-YVZJFKFKSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tong Zhang et al.
Analytical chemistry, 84(4), 1994-2001 (2012-03-14)
In this study, we assessed three liquid chromatographic platforms: reversed phase (RP), aqueous normal phase (ANP), and hydrophilic interaction (HILIC) for the analysis of polar metabolite standard mixtures and for their coverage of urinary metabolites. The two zwitterionic HILIC columns
Wen Shan Yew et al.
Biochemistry, 45(49), 14582-14597 (2006-12-06)
Many members of the mechanistically diverse enolase superfamily have unknown functions. In this report we use both genome (operon) context and screening of a library of acid sugars to assign the L-fuconate dehydratase (FucD) function to a member of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service