Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49010

Sigma-Aldrich

Trichloroacetic acid solution

6.1 N (approx. 100% w/v)

Synonym(s):

TCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
Beilstein/REAXYS Number:
970119
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

liquid

Quality Level

concentration

6.1 N (approx. 100% w/v)

SMILES string

OC(=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Trichloroacetic acid solution is traditionally used to precipitate protein. It can be used to determine protein concentration by quantitative precipitation. Trichloroacetic acid can also be used as a decalcifier and fixative in microscopy.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Caio A C G Brunharo et al.
Journal of agricultural and food chemistry, 67(31), 8431-8440 (2019-05-09)
Glufosinate-resistant Lolium perenne L. spp. multiflorum biotypes from Oregon exhibited resistance levels up to 2.8-fold the field rate. One resistant biotype (MG) had an amino acid substitution in glutamine synthetase 2 (GS2), whereas the other (OR) exhibited the wild-type genotype.
Ying Wang et al.
Diabetes, 63(8), 2643-2655 (2014-03-13)
After diabetes, the heart has a singular reliance on fatty acid (FA) for energy production, which is achieved by increased coronary lipoprotein lipase (LPL) that breaks down circulating triglycerides. Coronary LPL originates from cardiomyocytes, and to translocate to the vascular
Inga Müller et al.
PloS one, 9(5), e97285-e97285 (2014-05-27)
Members of the MYC family are the most frequently deregulated oncogenes in human cancer and are often correlated with aggressive disease and/or poorly differentiated tumors. Since patients with MYCN-amplified neuroblastoma have a poor prognosis, targeting MYCN using small molecule inhibitors
Hikmat Al-Ahmadie et al.
Cancer discovery, 4(9), 1014-1021 (2014-06-18)
Metastatic solid tumors are almost invariably fatal. Patients with disseminated small-cell cancers have a particularly unfavorable prognosis, with most succumbing to their disease within two years. Here, we report on the genetic and functional analysis of an outlier curative response
Jui-Ling Hsu et al.
Naunyn-Schmiedeberg's archives of pharmacology, 387(10), 979-990 (2014-07-10)
Hormone-refractory metastatic prostate cancer (HRMPC), which is metastatic and resistant to hormone therapy, is an intractable problem in clinical treatment. Anthraquinone-based natural products and synthetic compounds have shown anticancer activity. However, cardiac toxicity is a major adverse reaction in these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service