Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44209

Sigma-Aldrich

Phytanoyl-Coenzyme A triethylammonium salt

≥95.0% (HPLC)

Synonym(s):

Coenzyme A S-(3,7,11,15-tetramethylhexadecanoate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H74N7O17P3S · C6H15N
Molecular Weight:
1163.24
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

assay

≥95.0% (HPLC)

form

powder

color

faint beige to faint brown
white to faint yellow

storage temp.

−20°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC(C)CC(SCCNC(CCNC([C@H](O)C(C)(C)COP(OP(OC[C@H]1O[C@@H](N2C=NC3=C(N)N=CN=C32)[C@H](O)[C@@H]1OP(O)(O)=O)(O)=O)(O)=O)=O)=O)=O.CCN(CC)CC

InChI

1S/C41H74N7O17P3S.C6H15N/c1-26(2)11-8-12-27(3)13-9-14-28(4)15-10-16-29(5)21-32(50)69-20-19-43-31(49)17-18-44-39(53)36(52)41(6,7)23-62-68(59,60)65-67(57,58)61-22-30-35(64-66(54,55)56)34(51)40(63-30)48-25-47-33-37(42)45-24-46-38(33)48;1-4-7(5-2)6-3/h24-30,34-36,40,51-52H,8-23H2,1-7H3,(H,43,49)(H,44,53)(H,57,58)(H,59,60)(H2,42,45,46)(H2,54,55,56);4-6H2,1-3H3/t27?,28?,29?,30-,34-,35-,36+,40-;/m1./s1

InChI key

DZYNYSLSFBTWSM-IDHGLTIUSA-N

General description

Phytanoyl-Coenzyme A is an activated form of phytanic acid during phytanic acid α-oxidation. Phytanic acid is a derivative of phytol, which is a side chain of chlorophyll.[1]

Biochem/physiol Actions

Phytanoyl-Coenzyme A (phytanoyl-Co A) enters the peroxisomes during phytanic acid α-oxidation. It is converted to 2-hydroxyphytanoyl-CoA by the enzyme phytanoyl-CoA hydroxylase.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human metabolism of phytanic acid and pristanic acid
Verhoeven N M and Jakobs C
Progress in Lipid Research, 40(6), 453-466 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service