Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41563

Sigma-Aldrich

Manganese Peroxidase from Nematoloma frowardii

≥4.2 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

biological source

fungus (Nematoloma frowardii)

form

powder

specific activity

≥4.2 U/mg

storage temp.

−20°C

Application

Manganese peroxidase (MnP) is used to oxidize Mn2+ to Mn3+ in the presence of hydrogen peroxide. It is used for the biodegreadation of macromolecular substances such as lignin and humic substances [1].

Biochem/physiol Actions

Manganese peroxidase (MnP) is a hemecontaining glycoprotein that is produced by ligninolytic basidiomycetes. It requires hydrogen peroxide as an oxidant. MnP oxidizes Mn2+ to Mn3+. Mn3+ oxidizes phenolic rings to phenoxy radicals which results in the decomposition of various compounds [1].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which oxidizes 1 μmol Mn2+ to Mn3+ per minute at pH 4.5 and 25°C (in the presence of H2O2).

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hofrichter et al.
Applied and environmental microbiology, 64(2), 399-404 (2005-12-14)
The direct involvement of manganese peroxidase (MnP) in the mineralization of natural and xenobiotic compounds was evaluated. A broad spectrum of aromatic substances were partially mineralized by the MnP system of the white rot fungus Nematoloma frowardii. The cell-free MnP
Tatsuki Sugiura et al.
FEMS microbiology letters, 331(1), 81-88 (2012-04-18)
We identified a highly expressed protein (BUNA2) by two-dimensional gel electrophoresis from the hyper lignin-degrading fungus Phanerochaete sordida YK-624 under wood-rotting conditions. Partial amino acid sequences of BUNA2 were determined by LC-MS/MS analysis, and BUNA2 gene (bee2) and promoter region
Hiroshi Nishimura et al.
Organic & biomolecular chemistry, 10(31), 6432-6442 (2012-06-29)
New ceriporic acids-alkadienyl and alkenyl itaconic acids having a bis-allyl (3-[(Z,Z)-hexadec-7,10-dienyl]-itaconic acid; ceriporic acid G) and a monoene (3-[(Z)-octadec-9-enyl]-itaconic acid; ceriporic acid H) structure in their side chains-were isolated from the cultures of the selective lignin-degrading fungus Ceriporiopsis subvermispora. The
Guanyu Zheng et al.
Bioresource technology, 126, 397-403 (2012-04-24)
A novel approach was developed using oil-in-water (O/W) microemulsions formed with non-ionic surfactant, cosurfactant (1-pentanol) and linseed oil, at the cosurfactant to surfactant ratio (C/S ratio, w/w) of 1:3 and oil to surfactant ratio (O/S ratio, w/w) of 1:10, to
Tomer M Salame et al.
Applied and environmental microbiology, 78(15), 5341-5352 (2012-05-29)
Pleurotus ostreatus (the oyster mushroom) and other white rot filamentous basidiomycetes are key players in the global carbon cycle. P. ostreatus is also a commercially important edible fungus with medicinal properties and is important for biotechnological and environmental applications. Efficient

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service