Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38127

Sigma-Aldrich

AZCL-Galactan

substrate for galactanase

Synonym(s):

Azurine-cross-linked galactan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

solid

storage temp.

2-8°C

Application

Azurine-crosslinked-potato galactan, which has been purified from potato fibre and treated to remove most of the arabino- furanosyl residues. This substrate is useful for the measurement of endo-1,4-β-D-galactanase in crude enzyme mixtures.

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hildegard Watzlawick
Bio-protocol, 7(7), e2206-e2206 (2017-04-05)
The activity of the endo-β-1,4-galactanase GanB from B. subtilis on the high molecular weight β-1,4-galactan was determined quantitatively by the measurement of the increase of the reducing power or with the dyed substrate Azo-galactan. The generated degradation products were analyzed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service