Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

274429

Sigma-Aldrich

Methyl cellulose

average Mn ~40,000, powder, viscosity: 400 cP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

mol wt

average Mn ~40,000

extent of labeling

1.6-1.9 mol methoxy per mol cellulose (D.S.)
27.5-31.5 wt. % methoxy

surface tension

53-59 dyn/cm, 25 °C, 0.05 wt. %

viscosity

300-560 cP, 2 % in water(20)

transition temp

gel point 50-55 °C

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thickener for aqueous and non-aqueous systems, clear films with grease resistance, binders, lubricants, steric stabilizer and water retention aid.

Features and Benefits

Dissolves in water, undergoes reversible gelation upon heating, non-ionic, does not complex with ionic species and is surface active and enzyme resistant. Solutions are pseudoplastic.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rainelli Koumangoye et al.
Cellular and molecular gastroenterology and hepatology, 9(2), 239-255 (2019-10-28)
Infections resulting from intestinal yeast and bacteria affect a large number of patients with deficits in absorptive or secretory epithelial transport mechanisms. The basolateral Na+-K+-2Cl- cotransporter (NKCC1) has been implicated in intestinal epithelial fluid secretion. Two patients with deleterious heterozygous
Fanny Matheis et al.
Cell, 180(1), 64-78 (2020-01-11)
Enteric-associated neurons (EANs) are closely associated with immune cells and continuously monitor and modulate homeostatic intestinal functions, including motility and nutrient sensing. Bidirectional interactions between neuronal and immune cells are altered during disease processes such as neurodegeneration or irritable bowel
Ram Dixit et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(2), 492-497 (2009-01-08)
Microtubules are polarized polymers that exhibit dynamic instability, with alternating phases of elongation and shortening, particularly at the more dynamic plus-end. Microtubule plus-end tracking proteins (+TIPs) localize to and track with growing microtubule plus-ends in the cell. +TIPs regulate microtubule
Elsa-Noah N'Diaye et al.
EMBO reports, 10(2), 173-179 (2009-01-17)
Ubiquilins (UBQLNs) are adaptor proteins thought to deliver ubiquitinated substrates to proteasomes. Here, we show a role for UBQLN in autophagy: enforced expression of UBQLN protects cells from starvation-induced death, whereas depletion of UBQLN renders cells more susceptible. The UBQLN
Fumiyo Takabayashi et al.
The British journal of nutrition, 110(7), 1195-1200 (2013-02-26)
We have observed a stiffening of the gastric wall in mice following oral administration of viscous methyl cellulose (MC) solution as a vehicle for food chemicals, an effect which has not previously been reported. To further investigate this effect, young

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service