Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16321

Sigma-Aldrich

Tridecyl β-D-maltoside

≥99.0% (TLC)

Synonym(s):

n-Tridecyl β-D-maltopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H48O11
CAS Number:
Molecular Weight:
524.64
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

assay

≥99.0% (TLC)

form

solid

technique(s)

electrophoresis: suitable
protein purification: suitable
protein quantification: suitable

solubility

H2O: 10 mg/mL, clear, colorless

SMILES string

CCCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C25H48O11/c1-2-3-4-5-6-7-8-9-10-11-12-13-33-24-22(32)20(30)23(17(15-27)35-24)36-25-21(31)19(29)18(28)16(14-26)34-25/h16-32H,2-15H2,1H3/t16-,17-,18-,19+,20-,21-,22-,23-,24-,25-/m1/s1

Inchi Key

JZPRPDMVGIRGBV-IYBATYGCSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel A P Gutmann et al.
Protein science : a publication of the Protein Society, 16(7), 1422-1428 (2007-06-15)
One key to successful crystallization of membrane proteins is the identification of detergents that maintain the protein in a soluble, monodispersed state. Because of their hydrophobic nature, membrane proteins are particularly prone to forming insoluble aggregates over time. This nonspecific
null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service