Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16202

Supelco

Amitraz-(N-methyl-d3)

PESTANAL®, analytical standard

Synonym(s):

N′-(2,4-Dimethylphenyl)-N-{[(2,4-dimethylphenyl)imino]methyl}-N-(methyl-d3)methanimidamide, Amitraz-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D3H20N3
Molecular Weight:
296.42
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

isotopic purity

≥95.0 atom % D

product line

PESTANAL®

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

83-87 °C

application(s)

agriculture

format

neat

SMILES string

CC1=CC(C)=CC=C1/N=C/N(C([2H])([2H])[2H])/C=N/C2=C(C)C=C(C)C=C2

General description

Amitraz-(N-methyl-d3) is an isotope labeled analog of the acaricide and insecticide amitraz, wherein N-methyl protons of amitraz are replaced by deuterium.[1]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amitraz poisoning in children
Yaramis A, et al.
Human & Experimental Toxicology, 19(8), 431-433 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service