Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15760

Sigma-Aldrich

Boron trifluoride acetic acid complex

technical, ~36% BF3 basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · 2CH3COOH
CAS Number:
Molecular Weight:
187.91
Beilstein/REAXYS Number:
3686517
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

technical

form

solid

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

~36% (BF3)

density

1.353 g/mL at 20 °C (lit.)

SMILES string

CC(O)=O.CC(O)=O.FB(F)F

InChI

1S/2C2H4O2.BF3/c2*1-2(3)4;2-1(3)4/h2*1H3,(H,3,4);

InChI key

COTMJGCQSLZICX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

181.4 °F - closed cup

flash_point_c

83 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service